Assay Design
Gene Transcripts
- Gene Symbol
- ncm
- Entrez Gene ID
-
35099
- Gene Name
- nucampholin
- Gene Aliases
-
CG12750, Dmel\CG12750, Dmel_CG12750, E-2a, NCM, l(2)SH0931, l(2)SH2 0931
- Chromosome Location
-
Chr.2L: 18444507 - 18448957 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0086707
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136033.3 |
NP_609877.2 |
1 - 2 |
710 |
81 |
GenBank mRNA |
BT100034.1 |
- |
1 - 2 |
680 |
81 |
View Full Product Details