Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274744.2 | NP_001261673.1 | 3 - 4 | 1955 | 69 |
NM_001274745.1 | NP_001261674.1 | 3 - 4 | 1955 | 69 | |
NM_001274746.1 | NP_001261675.1 | 3 - 4 | 1955 | 69 | |
NM_001300089.2 | NP_001287018.1 | 3 - 4 | 1955 | 69 | |
NM_144182.2 | NP_652439.1 | 3 - 4 | 1955 | 69 | |
GenBank mRNA | AY069421.1 | - | 3 - 4 | 1987 | 69 |