Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001273787.1 | NP_001260716.1 | 2 - 3 | 1619 | 123 |
NM_136342.2 | NP_610186.2 | 2 - 3 | 1619 | 123 | |
NM_144205.5 | NP_652462.3 | 3 - 4 | 1650 | 123 | |
GenBank mRNA | BT029948.1 | - | 1 - 2 | 780 | 123 |