Assay Design
Gene Transcripts
- Gene Symbol
- ACXE
- Entrez Gene ID
-
53426
- Gene Name
- Adenylyl cyclase X E
- Gene Aliases
-
AC, CG17178, DACXE, Dmel\CG17178, Dmel_CG17178
- Chromosome Location
-
Chr.2L: 12925611 - 12929823 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040506
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144344.3 |
NP_652601.3 |
5 - 6 |
1253 |
75 |
View Full Product Details