Assay Design



Gene Transcripts

Gene Symbol
HGTX
Entrez Gene ID
53446
Gene Name
CG13475-PA
Gene Aliases
CG13475, CG4745, Dmel\CG13475, Dmel_CG13475, Nk6, Nkx6, dNkx6, nkx6
Chromosome Location
Chr.3L: 14583222 - 14599540 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0040318
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382022.1 NP_001368954.1 1 - 2 1759 57
NM_144357.3 NP_652614.1 1 - 2 1759 57
GenBank mRNA AF220236.2 - 1 - 2 1787 57
BT015992.1 - 1 - 2 1756 57
BT022198.1 - 1 - 2 1964 57

View Full Product Details