Assay Design
Gene Transcripts
- Gene Symbol
- lectin-46Ca
- Entrez Gene ID
-
53523
- Gene Name
- CG1656-PA
- Gene Aliases
-
AC 007330A, AC007330a, CG1656, Dmel\CG1656, Dmel_CG1656, Lectin-46CA, Lectin-46Ca
- Chromosome Location
-
Chr.2R: 9817300 - 9818535 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040093
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144376.2 |
NP_652633.1 |
1 - 2 |
175 |
64 |
View Full Product Details