Assay Design



Gene Transcripts

Gene Symbol
mam
Entrez Gene ID
36555
Gene Name
mastermind
Gene Aliases
CG8118, DMam, Dm0064, Dmel\CG8118, Dmel_CG8118, MAM, Mam, MamH, N-2G, SR2-4, dMam, l(2)03505, l(2)04615, mam(CAG)[[7]]
Chromosome Location
Chr.2R: 13991203 - 14060356 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0002643
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299472.1 NP_001286401.1 4 - 5 3712 61
NM_080376.2 NP_525115.1 5 - 6 3795 61
NM_166030.2 NP_725354.1 4 - 5 3418 61

View Full Product Details