Assay Design



Gene Transcripts

Gene Symbol
ave
Entrez Gene ID
246638
Gene Name
aveugle
Gene Aliases
BcDNA:RE67675, CG30476, CG30476/Ave, Dmel\CG30476, Dmel_CG30476, HYP, hyp
Chromosome Location
Chr.2R: 14752521 - 14753269 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0050476
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_166066.3 NP_725413.1 1 - 2 225 86
GenBank mRNA BT001710.1 - 1 - 2 220 86
KX531281.1 - 1 - 2 156 86

View Full Product Details