Assay Design
Gene Transcripts
- Gene Symbol
- Obp51a
- Entrez Gene ID
-
246666
- Gene Name
- Odorant-binding protein 51a
- Gene Aliases
-
51a, CG30473, DmelObp51a, Dmel\CG30473, Dmel_CG30473, Obp-c, anon-AE003812.1
- Chromosome Location
-
Chr.2R: 15024375 - 15024839 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043530
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166079.2 |
NP_725436.1 |
1 - 2 |
60 |
106 |
GenBank mRNA |
AF457145.1 |
- |
1 - 2 |
60 |
106 |
View Full Product Details