Assay Design



Gene Transcripts

Gene Symbol
csul
Entrez Gene ID
36809
Gene Name
capsuleen
Gene Aliases
CG3730, Csul, DART5, Dart5, Dmel\CG3730, Dmel_CG3730, JBP1, PRMT5, dPRMT5, dart5, dprmt5
Chromosome Location
Chr.2R: 16264652 - 16266959 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015925
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057836.4 NP_477184.1 4 - 5 1257 69
NM_166158.2 NP_725552.1 4 - 5 1242 69
GenBank mRNA AY122153.1 - 4 - 5 1225 69

View Full Product Details