Assay Design



Gene Transcripts

Gene Symbol
Lmpt
Entrez Gene ID
39889
Gene Name
Limpet
Gene Aliases
BcDNA:RE37250, CG11914, CG11915, CG11916, CG32171, CG33256, CG42679, CG9959, Dmel\CG42679, Dmel_CG42679, lmpt
Chromosome Location
Chr.3L: 16865295 - 16922487 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261565
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169985.1 NP_001163456.1 3 - 4 580 63
NM_001202193.2 NP_001189122.1 5 - 6 3267 63
NM_001259879.2 NP_001246808.1 8 - 9 1671 63
NM_001275047.1 NP_001261976.2 6 - 7 1456 63
NM_001316485.1 NP_001303414.1 4 - 5 660 63
NM_140673.5 NP_648930.1 8 - 9 1682 63
NM_140676.4 NP_648933.3 3 - 4 576 63
NM_168694.3 NP_730213.1 4 - 5 865 63
NM_170635.3 NP_730212.1 7 - 8 1626 63
NM_206387.3 NP_996109.1 8 - 9 1570 63
NM_206388.3 NP_996110.1 8 - 9 1794 63
NM_206389.3 NP_996111.1 4 - 5 876 63
NM_206390.2 NP_996112.1 3 - 4 598 63
GenBank mRNA AJ418713.1 - 8 - 9 1664 63
AY069696.1 - 8 - 9 1664 63
AY071605.1 - 3 - 4 580 63
AY128470.1 - 3 - 4 598 63
BT009969.1 - 8 - 9 1679 63
BT044112.1 - 4 - 5 864 63
BT100317.1 - 7 - 8 1624 63

View Full Product Details