Assay Design



Gene Transcripts

Gene Symbol
Lmpt
Entrez Gene ID
39889
Gene Name
Limpet
Gene Aliases
BcDNA:RE37250, CG11914, CG11915, CG11916, CG32171, CG33256, CG42679, CG9959, Dmel\CG42679, Dmel_CG42679, lmpt
Chromosome Location
Chr.3L: 16865295 - 16922487 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261565
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259879.2 NP_001246808.1 3 - 4 500 68
NM_140673.5 NP_648930.1 3 - 4 511 68
NM_206388.3 NP_996110.1 3 - 4 623 68
GenBank mRNA AJ418713.1 - 3 - 4 493 68
AY069696.1 - 3 - 4 493 68
BT009969.1 - 3 - 4 508 68

View Full Product Details