Assay Design
Gene Transcripts
- Gene Symbol
- dec-1
- Entrez Gene ID
-
31691
- Gene Name
- defective chorion 1
- Gene Aliases
-
39051, CG2175, Dmel\CG2175, Dmel_CG2175, Fc, Fc1, Fcp7C, dec, dec1, dec[[1]], fs(1)11-549, fs(1)12-1873, fs(1)12-2514, fs(1)12-3512, fs(1)12-365, fs(1)12-3907, fs(1)12-403, fs(1)12-4860, fs(1)13-453, fs(1)13C-57, fs(1)13C-73, fs(1)14-963, fs(1)14A-114, fs(1)14E19, fs(1)1501, fs(1)384, fs(1)5, fs(1)A257, fs(1)A384, fs(1)C1, fs(1)M102, fs(1)M1021
- Chromosome Location
-
Chr.X: 7867161 - 7873222 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000427
View Full Product Details