Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001298192.1 | NP_001285121.1 | 2 - 3 | 430 | 66 |
NM_001298193.1 | NP_001285122.1 | 2 - 3 | 341 | 66 | |
NM_001298194.1 | NP_001285123.1 | 1 - 2 | 502 | 66 | |
NM_078568.3 | NP_511123.2 | 2 - 3 | 336 | 66 | |
NM_167284.2 | NP_727525.1 | 2 - 3 | 418 | 66 | |
NM_206685.2 | NP_996408.1 | 2 - 3 | 324 | 66 | |
GenBank mRNA | BT023772.1 | - | 2 - 3 | 395 | 66 |