Assay Design



Gene Transcripts

Gene Symbol
Ptp10D
Entrez Gene ID
32115
Gene Name
Protein tyrosine phosphatase 10D
Gene Aliases
10D, CG1817, CT4920, DPTP, DPTP10D, DPTP[[10D]], Dmel\CG1817, Dmel_CG1817, PTP10D, Ptp10, R-PTP 10D, RPTP10d, dptp, ptp10D, ptp10d
Chromosome Location
Chr.X: 11622015 - 11677338 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004370
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144715.2 NP_001138187.1 2 - 3 734 84
NM_001272524.1 NP_001259453.1 2 - 3 562 84
NM_001272525.1 NP_001259454.1 3 - 4 607 84
NM_167292.3 NP_727544.2 2 - 3 562 84
NM_206690.2 NP_996413.2 2 - 3 639 84
NM_206691.3 NP_996414.2 2 - 3 811 84

View Full Product Details