Assay Design



Gene Transcripts

Gene Symbol
yl
Entrez Gene ID
32367
Gene Name
yolkless
Gene Aliases
CG1372, CT3114, DmYPR, Dmel\CG1372, Dmel_CG1372, LR13, YL, Yl, fs(1)148, fs(1)5, fs(1)A148, fs(1)K184, fs(1)L186, fs(1)L193, fs(1)L196, fs(1)L211, fs(1)M205, fs(1)M25, fs(1)yl, fs(1)yolkless
Chromosome Location
Chr.X: 14193214 - 14199958 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004649
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078596.3 NP_511151.2 3 - 4 3501 69
NM_206710.2 NP_996433.1 3 - 4 3501 69
GenBank mRNA BT150470.1 - 3 - 4 3496 69

View Full Product Details