Assay Design



Gene Transcripts

Gene Symbol
Pex12
Entrez Gene ID
33256
Gene Name
Peroxin 12
Gene Aliases
CG3639, DmelPex12, Dmel\CG3639, Dmel_CG3639, PEX12, pex12
Chromosome Location
Chr.2L: 815993 - 818096 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031282
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272915.1 NP_001259844.1 1 - 1 1260 83

View Full Product Details