Assay Design



Gene Transcripts

Gene Symbol
Tango14
Entrez Gene ID
33298
Gene Name
Transport and Golgi organization 14
Gene Aliases
BcDNA:SD10843, CG4775, Dmel\CG4775, Dmel_CG4775, TANGO14, Tango 14, l(2)k00619
Chromosome Location
Chr.2L: 1158661 - 1160220 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031312
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272928.1 NP_001259857.1 2 - 3 738 141
NM_134733.3 NP_608577.2 1 - 2 812 141
GenBank mRNA AY089679.1 - 1 - 2 794 141

View Full Product Details