Assay Design
Gene Transcripts
- Gene Symbol
- VGlut
- Entrez Gene ID
-
33427
- Gene Name
- Vesicular glutamate transporter
- Gene Aliases
-
CG9887, DV-Glut, DVGLUT, DVGluT, DVGlut, Dmel\CG9887, Dmel_CG9887, DvGluT, DvGlut, MFS11, VGLUT, VGluT, Vglut, dVGLUT, dVGlut, dvGluT, dvGlut, dvglut, vGLUT, vGluT, vGlut, vglut
- Chromosome Location
-
Chr.2L: 2400295 - 2410662 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031424
View Full Product Details