Assay Design



Gene Transcripts

Gene Symbol
Drp1
Entrez Gene ID
33445
Gene Name
Dynamin related protein 1
Gene Aliases
CG3210, DNM1L, DRP, DRP1, Dmel\CG3210, Dmel_CG3210, Dnm1/Drp1, Drp, Drp-1, drp, drp-1, drp1, frat, l(2)22Fc, l(2)22Fd, l(2)ND2, l(2)ND3
Chromosome Location
Chr.2L: 2581281 - 2585430 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026479
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273017.1 NP_001259946.1 3 - 4 1068 72
NM_134850.3 NP_608694.2 3 - 4 1068 72
GenBank mRNA BT133244.1 - 3 - 4 1086 72

View Full Product Details