Assay Design
Gene Transcripts
- Gene Symbol
- Drp1
- Entrez Gene ID
-
33445
- Gene Name
- Dynamin related protein 1
- Gene Aliases
-
CG3210, DNM1L, DRP, DRP1, Dmel\CG3210, Dmel_CG3210, Dnm1/Drp1, Drp, Drp-1, drp, drp-1, drp1, frat, l(2)22Fc, l(2)22Fd, l(2)ND2, l(2)ND3
- Chromosome Location
-
Chr.2L: 2581281 - 2585430 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0026479
View Full Product Details