Assay Design



Gene Transcripts

Gene Symbol
E23
Entrez Gene ID
43895
Gene Name
Early gene at 23
Gene Aliases
CG15410, CG3327, Dmel\CG3327, Dmel_CG3327
Chromosome Location
Chr.2L: 3334875 - 3354829 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020445
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079909.3 NP_524648.3 6 - 7 3002 77
NM_134924.2 NP_608768.2 6 - 7 3754 77
NM_205900.2 NP_995622.1 6 - 7 3119 77
GenBank mRNA BT133432.1 - 6 - 7 3119 77

View Full Product Details