Assay Design



Gene Transcripts

Gene Symbol
E23
Entrez Gene ID
43895
Gene Name
Early gene at 23
Gene Aliases
CG15410, CG3327, Dmel\CG3327, Dmel_CG3327
Chromosome Location
Chr.2L: 3334875 - 3354829 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0020445
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079909.3 NP_524648.3 8 - 9 3412 102
NM_134924.2 NP_608768.2 8 - 9 4164 102
NM_205900.2 NP_995622.1 8 - 9 3529 102
GenBank mRNA BT133432.1 - 8 - 9 3529 102

View Full Product Details