Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169928.2 | NP_001163399.1 | 6 - 7 | 1316 | 108 |
NM_001169929.2 | NP_001163400.1 | 6 - 7 | 1296 | 108 | |
NM_140043.5 | NP_648300.3 | 7 - 8 | 1302 | 108 | |
NM_168325.4 | NP_729469.2 | 7 - 8 | 1322 | 108 | |
GenBank mRNA | AY069422.1 | - | 7 - 8 | 1178 | 108 |
BT003777.1 | - | 7 - 8 | 1237 | 108 | |
BT125863.1 | - | 6 - 7 | 1208 | 108 | |
BT133238.1 | - | 6 - 7 | 1194 | 108 |