Assay Design
Gene Transcripts
- Gene Symbol
- Or67d
- Entrez Gene ID
-
39191
- Gene Name
- Odorant receptor 67d
- Gene Aliases
-
67d, CG14157, DmOr67d, DmelOr67d, Dmel\CG14157, Dmel_CG14157, OR67d
- Chromosome Location
-
Chr.3L: 10273204 - 10274624 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036080
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140133.2 |
NP_648390.2 |
3 - 4 |
960 |
98 |
View Full Product Details