Assay Design



Gene Transcripts

Gene Symbol
rols
Entrez Gene ID
39368
Gene Name
rolling pebbles
Gene Aliases
ANTS, Ants, CG12277, CG17155, CG32096, CG5679, Dmel\CG32096, Dmel_CG32096, Rol/Ants, Rols, Rols6, Rols7, anon-WO0257455.19, ants, ants/rols, l(3)08232, l(3)68Fd, l(3)j2A6, rols\ants
Chromosome Location
Chr.3L: 12008225 - 12064859 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041096
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274807.1 NP_001261736.1 2 - 3 793 85
NM_001274809.1 NP_001261738.1 2 - 3 752 85
NM_140285.3 NP_648542.1 1 - 2 1273 85
NM_168488.2 NP_729779.1 2 - 3 752 85
NM_168489.2 NP_729780.1 2 - 3 776 85
NM_168490.2 NP_729781.1 2 - 3 793 85
GenBank mRNA AF440568.1 - 1 - 2 349 85
AF440569.1 - 1 - 2 349 85
AY058388.1 - 1 - 2 349 85

View Full Product Details