Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274807.1 | NP_001261736.1 | 7 - 8 | 5252 | 64 |
NM_001274808.1 | NP_001261737.1 | 6 - 7 | 5378 | 64 | |
NM_001274809.1 | NP_001261738.1 | 6 - 7 | 4908 | 64 | |
NM_001274810.1 | NP_001261739.1 | 6 - 7 | 5375 | 64 | |
NM_140285.3 | NP_648542.1 | 5 - 6 | 5432 | 64 | |
NM_168487.2 | NP_729778.1 | 7 - 8 | 5681 | 64 | |
NM_168488.2 | NP_729779.1 | 6 - 7 | 4911 | 64 | |
NM_168489.2 | NP_729780.1 | 7 - 8 | 5238 | 64 | |
NM_168490.2 | NP_729781.1 | 7 - 8 | 5255 | 64 | |
GenBank mRNA | AF440568.1 | - | 6 - 7 | 4811 | 64 |
AF440569.1 | - | 5 - 6 | 4508 | 64 | |
AY058388.1 | - | 5 - 6 | 4508 | 64 |