Assay Design
Gene Transcripts
- Gene Symbol
- Smyd4-2
- Entrez Gene ID
-
39414
- Gene Name
- SET and MYND domain containing, class 4, member 2
- Gene Aliases
-
CG14122, Dmel\CG14122, Dmel_CG14122, Smyd4, dSmyd4
- Chromosome Location
-
Chr.3L: 12399539 - 12401935 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036282
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140317.3 |
NP_648574.1 |
4 - 5 |
1792 |
102 |
GenBank mRNA |
AY071300.1 |
- |
4 - 5 |
1679 |
102 |
View Full Product Details