Assay Design



Gene Transcripts

Gene Symbol
x16
Entrez Gene ID
33967
Gene Name
x16 splicing factor
Gene Aliases
2/30, 9G8, CG10203, Dmel\CG10203, Dmel_CG10203, Dx16, Dxl6, RBP1-like, TN166, X16, X16/9G8, XL6, Xl6, anon-WO0118547.78, d9G8, dx16, dxl6, l(2)k00230, xl6
Chromosome Location
Chr.2L: 6914301 - 6920210 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028554
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_164718.2 NP_723226.1 1 - 2 690 67
GenBank mRNA AF232774.1 - 1 - 2 645 67
AJ249466.1 - 1 - 2 658 67
AY095057.1 - 1 - 2 653 67

View Full Product Details