Assay Design
Gene Transcripts
- Gene Symbol
- LanB1
- Entrez Gene ID
-
34068
- Gene Name
- CG7123-PA
- Gene Aliases
-
BcDNA:LD21794, CG7123, CT22015, Dmel\CG7123, Dmel_CG7123, EP-600, LM-B1/beta, LMbeta1, Lam-B1, LamB1, Lambeta, Lan B1, LanB, beta1, lamB1, laminin, lanB1, mspl
- Chromosome Location
-
Chr.2L: 7811438 - 7820771 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261800
View Full Product Details