Assay Design



Gene Transcripts

Gene Symbol
LanB1
Entrez Gene ID
34068
Gene Name
CG7123-PA
Gene Aliases
BcDNA:LD21794, CG7123, CT22015, Dmel\CG7123, Dmel_CG7123, EP-600, LM-B1/beta, LMbeta1, Lam-B1, LamB1, Lambeta, Lan B1, LanB, beta1, lamB1, laminin, lanB1, mspl
Chromosome Location
Chr.2L: 7811438 - 7820771 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261800
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057270.5 NP_476618.1 4 - 5 5930 142
NM_164773.3 NP_723319.1 4 - 5 5642 142

View Full Product Details