Assay Design



Gene Transcripts

Gene Symbol
Piezo
Entrez Gene ID
34112
Gene Name
CG44122-PE
Gene Aliases
CG18103, CG31608, CG44122, CG8486, DmPIEZO, DmPiezo, Dmel\CG44122, Dmel_CG44122, Dmpiezo, OrfKD, anon-EST:Liang-1.32, clone 1.32, fos28F, fos500, piezo
Chromosome Location
Chr.2L: 8163073 - 8190137 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264953
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042881.5 NP_001036346.3 16 - 17 4977 89
NM_001169436.3 NP_001162907.1 15 - 16 4755 89
NM_001201789.2 NP_001188718.1 16 - 17 4812 89
NM_001201790.2 NP_001188719.1 15 - 16 4755 89
NM_001273297.1 NP_001260226.1 17 - 18 5034 89
NM_001273299.1 NP_001260228.1 15 - 16 4755 89
NM_135344.6 NP_609188.4 16 - 17 4751 89
NM_164795.6 NP_723354.3 16 - 17 4977 89

View Full Product Details