Assay Design



Gene Transcripts

Gene Symbol
rost
Entrez Gene ID
34222
Gene Name
rolling stone
Gene Aliases
CG9552, Dmel\CG9552, Dmel_CG9552
Chromosome Location
Chr.2L: 8985936 - 8989539 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011705
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273345.1 NP_001260274.1 3 - 4 838 64
NM_001273346.1 NP_001260275.1 2 - 3 708 64
NM_164842.2 NP_723435.1 2 - 3 603 64
GenBank mRNA BT016130.1 - 2 - 3 580 64

View Full Product Details