Assay Design



Gene Transcripts

Gene Symbol
Oatp30B
Entrez Gene ID
34268
Gene Name
Organic anion transporting polypeptide 30B
Gene Aliases
CG3811, Dmel\CG3811, Dmel_CG3811, oatp 30B, oatp30B
Chromosome Location
Chr.2L: 9521210 - 9540060 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032123
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135449.4 NP_609293.2 8 - 9 3223 72
NM_164857.3 NP_723463.1 8 - 9 3278 72
NM_205945.2 NP_995667.1 6 - 7 2619 72
NM_205946.2 NP_995668.1 7 - 8 3097 72

View Full Product Details