Assay Design



Gene Transcripts

Gene Symbol
RluA-1
Entrez Gene ID
34438
Gene Name
RluA pseudouridine synthase 1
Gene Aliases
CG13143, CG31719, CG7475, Dmel\CG31719, Dmel_CG31719, RluA1
Chromosome Location
Chr.2L: 10463340 - 10474349 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051719
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_164930.2 NP_723592.1 5 - 6 1485 132
NM_164931.2 NP_723593.1 5 - 6 1619 132
NM_164932.2 NP_723594.1 5 - 6 1326 132
GenBank mRNA BT044578.1 - 5 - 6 1484 132

View Full Product Details