Assay Design



Gene Transcripts

Gene Symbol
escl
Entrez Gene ID
34611
Gene Name
CG5202-PA
Gene Aliases
CG5202, CG5202-PA, Dmel\CG5202, Dmel_CG5202, ESC, ESC-LIKE, ESC1, ESCL
Chromosome Location
Chr.2L: 11990927 - 11992848 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032391
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298928.1 NP_001285857.1 2 - 3 540 68
NM_164979.2 NP_723702.1 2 - 3 540 68
GenBank mRNA BT015954.1 - 2 - 3 525 68

View Full Product Details