Assay Design



Gene Transcripts

Gene Symbol
JhI-21
Entrez Gene ID
34624
Gene Name
Juvenile hormone Inducible-21
Gene Aliases
CG12317, Dmel\CG12317, Dmel_CG12317, JHI-21, Jhl-21
Chromosome Location
Chr.2L: 12051755 - 12056042 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028425
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259067.1 NP_001245996.1 7 - 8 1397 76
NM_001298934.1 NP_001285863.1 7 - 8 1367 76
NM_001298935.1 NP_001285864.1 7 - 8 1430 76
NM_135698.4 NP_609542.1 7 - 8 1373 76
NM_164988.4 NP_723717.2 7 - 8 1337 76
GenBank mRNA AF273478.1 - 7 - 8 1335 76
AY051907.1 - 7 - 8 1322 76

View Full Product Details