Assay Design
Gene Transcripts
- Gene Symbol
- Cyp4ac2
- Entrez Gene ID
-
33755
- Gene Name
- CG17970-PB
- Gene Aliases
-
4ac2, CG17970, CYP4ac2, Dmel\CG17970, Dmel_CG17970
- Chromosome Location
-
Chr.2L: 5266392 - 5268455 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0031694
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135073.3 |
NP_608917.3 |
1 - 2 |
244 |
79 |
View Full Product Details