Assay Design



Gene Transcripts

Gene Symbol
mid
Entrez Gene ID
33770
Gene Name
midline
Gene Aliases
CG6634, Dmel\CG6634, Dmel_CG6634, H15, H15r, H15r/nmr2, MID, Mid, Mid(H15), TBX20, extra, los, nmr2
Chromosome Location
Chr.2L: 5461639 - 5467610 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261963
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001273167.2 NP_001260096.1 3 - 4 1197 95
NM_135083.4 NP_608927.2 3 - 4 1197 95
GenBank mRNA BT023769.1 - 3 - 4 1183 95

View Full Product Details