Assay Design



Gene Transcripts

Gene Symbol
CG11147
Entrez Gene ID
33803
Gene Name
CG11147-PA
Gene Aliases
Dmel\CG11147, Dmel_CG11147
Chromosome Location
Chr.2L: 5733675 - 5741244 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0031734
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135110.1 NP_608954.1 6 - 7 1407 97
NM_164658.2 NP_723114.1 5 - 6 1280 97
GenBank mRNA BT010210.1 - 6 - 7 1408 97

View Full Product Details