Assay Design



Gene Transcripts

Gene Symbol
Hsc70Cb
Entrez Gene ID
39557
Gene Name
CG6603-PA
Gene Aliases
0041/12, 0318/20, 1342/17, 1348/02, 1485/13, BEST:LD32772, CG6603, Dmel\CG6603, Dmel_CG6603, HSC-70, HSC70, HSC70cb, HSP110, HSP110-1, Hsc70, Hsp100, LD32772, anon-WO0118547.302, dHsp110, hsc70Cb, hsc70cb, l(3)00082, l(3)07621, l(3)134217, l(3)148513, l(3)70Ca, l(3)70Cb, l(3)S134217, l(3)S134802, l(3)S148513
Chromosome Location
Chr.3L: 14037019 - 14043686 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026418
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202172.2 NP_001189101.1 2 - 3 388 68
NM_001202173.2 NP_001189102.1 2 - 3 363 68
NM_001202174.2 NP_001189103.1 2 - 3 391 68
NM_001202175.2 NP_001189104.1 2 - 3 366 68
NM_001274893.1 NP_001261822.1 2 - 3 667 68
NM_140430.4 NP_648687.1 2 - 3 391 68
NM_168573.3 NP_729951.1 2 - 3 366 68
NM_168574.1 NP_729952.1 1 - 2 496 68
GenBank mRNA AY058632.1 - 2 - 3 342 68

View Full Product Details