Assay Design



Gene Transcripts

Gene Symbol
bmm
Entrez Gene ID
39611
Gene Name
brummer
Gene Aliases
ATGL, ATGL/bmm, Atgl, BMM, Bmm, CG5295, Dmel\CG5295, Dmel_CG5295, SCAP
Chromosome Location
Chr.3L: 14776496 - 14786412 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0036449
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169974.2 NP_001163445.1 4 - 5 1454 66
NM_140466.2 NP_648723.1 4 - 5 1454 66
GenBank mRNA AY051668.1 - 4 - 5 1399 66

View Full Product Details