Assay Design
Gene Transcripts
- Gene Symbol
- bmm
- Entrez Gene ID
-
39611
- Gene Name
- brummer
- Gene Aliases
-
ATGL, ATGL/bmm, Atgl, BMM, Bmm, CG5295, Dmel\CG5295, Dmel_CG5295, SCAP
- Chromosome Location
-
Chr.3L: 14776496 - 14786412 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036449
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140466.2 |
NP_648723.1 |
6 - 7 |
1804 |
59 |
GenBank mRNA |
AY051668.1 |
- |
6 - 7 |
1749 |
59 |
View Full Product Details