Assay Design



Gene Transcripts

Gene Symbol
elgi
Entrez Gene ID
39735
Gene Name
early girl
Gene Aliases
CG17033, Dmel\CG17033, Dmel_CG17033, Nrdp1, dNrdp1
Chromosome Location
Chr.3L: 15956550 - 15958433 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0283649
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274975.1 NP_001261904.1 3 - 4 796 82
NM_140559.1 NP_648816.1 3 - 4 873 82
GenBank mRNA AY051931.1 - 3 - 4 797 82

View Full Product Details