Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001274993.1 | NP_001261922.1 | 2 - 3 | 813 | 56 |
NM_140577.2 | NP_648834.1 | 2 - 3 | 414 | 56 | |
NM_168650.2 | NP_730105.3 | 2 - 3 | 813 | 56 | |
GenBank mRNA | BT133258.1 | - | 2 - 3 | 813 | 56 |