Assay Design



Gene Transcripts

Gene Symbol
Trs20
Entrez Gene ID
39769
Gene Name
TRAPP subunit 20
Gene Aliases
BcDNA:RE62842, CG5161, Dmel\CG5161, Dmel_CG5161, E(smoDN)B-left, Group B-left, TRAPP2C, TRAPPC2, Trs20p, l(3)72Dh
Chromosome Location
Chr.3L: 16131001 - 16131632 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266724
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_140584.3 NP_648841.1 1 - 2 144 70
GenBank mRNA AY071561.1 - 1 - 2 115 70
KX531681.1 - 1 - 2 94 70

View Full Product Details