Assay Design
Gene Transcripts
- Gene Symbol
- sinah
- Entrez Gene ID
-
39885
- Gene Name
- sina homologue
- Gene Aliases
-
CG13030, Dmel\CG13030, Dmel_CG13030, SinaH, anon-WO0140519.228, sina-h
- Chromosome Location
-
Chr.3L: 16859080 - 16860529 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259794
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140670.2 |
NP_648927.1 |
1 - 1 |
1239 |
79 |
GenBank mRNA |
BT044413.1 |
- |
1 - 1 |
1240 |
79 |
View Full Product Details