Assay Design
Gene Transcripts
- Gene Symbol
- Ir75b
- Entrez Gene ID
-
8673994
- Gene Name
- Ionotropic receptor 75b
- Gene Aliases
-
CG14586, CG14586-B, CG42643, DmelIR75b, Dmel\CG42643, Dmel_CG42643, IR75abc, IR75b, Ir75abc
- Chromosome Location
-
Chr.3L: 17818073 - 17820193 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261402
View Full Product Details