Assay Design
Gene Transcripts
- Gene Symbol
- Prestin
- Entrez Gene ID
-
39996
- Gene Name
- CG5485-PA
- Gene Aliases
-
CG5485, CG5845, Dmel\CG5485, Dmel_CG5485, Slc26a5, dPrestin, dpres, prestin
- Chromosome Location
-
Chr.3L: 17898889 - 17907812 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0036770
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_140767.2 |
NP_649024.1 |
3 - 4 |
1672 |
69 |
GenBank mRNA |
BT133214.1 |
- |
3 - 4 |
1640 |
69 |
View Full Product Details