Assay Design



Gene Transcripts

Gene Symbol
pdm2
Entrez Gene ID
34673
Gene Name
POU domain protein 2
Gene Aliases
CG12287, CG15486, CG15487, Dmel\CG12287, Dmel_CG12287, Miti, Oct-2, Oct2, Pdm, Pdm-2, Pdm2, dOct2, dPOU-28, dPOU28, dPOU28/pdm-2, dim, dpou28, miti, pdm, pdm-2
Chromosome Location
Chr.2L: 12657165 - 12686306 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004394
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298948.1 NP_001285877.1 2 - 3 1035 95
NM_001298949.1 NP_001285878.1 2 - 3 1592 95
NM_165017.2 NP_723763.1 2 - 3 933 95

View Full Product Details